Learn More
Bon Opus Biosciences Recombinant Mouse IL-21 -500ug
SDP
Supplier:  Bon Opus Biosciences BP111500UG
Encompass_Preferred
Recombinant Mouse IL-21 is produced by Human 293 Cells. The target gene encoding H18-S146 is expressed. Expression system: Human 293 Cells. Accession# Q9ES17. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP111
Catalog No. 50-208-0186