Learn More
Bon Opus Biosciences Recombinant Mouse IL-5 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CW7350UG
Encompass_Preferred
Recombinant Mouse Interleukin-5 is produced by our Mammalian expression system and the target gene encoding Met21-Gly133 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# P04401. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# CW73
Catalog No. 50-208-0454