Learn More
Bon Opus Biosciences Recombinant Mouse Insulin-2/Ins2 (C-His) -500ug
SDP
Catalog No. 502080156
Encompass_Preferred
Recombinant Mouse Insulin-2 is produced by Pichia pastoris. The target gene encoding F25-N110 is expressed with a 6His tag at the C terminus. Expression system: Pichia pastoris. Accession# P01326. Formulation: Supplied as a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP081
Catalog No. 50-208-0156 Supplier Bon Opus Biosciences Supplier No. BP081500UG
Recombinant Mouse Insulin-2 is produced by Pichia pastoris. The target gene encoding F25-N110 is expressed with a 6His tag at the C terminus. Expression system: Pichia pastoris. Accession# P01326. Formulation: Supplied as a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP081