Learn More
Bon Opus Biosciences Recombinant Mouse Interleukin-22/IL-22 (C-mFc) 500UG
SDP
Supplier:  Bon Opus Biosciences CM71500UG
Encompass_Preferred
Recombinant Mouse Interleukin-22 is produced by our Mammalian expression system and the target gene encoding Leu34-Val179 is expressed with a mFc tag at the C-terminus. Accession# Q9JJY9. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH7.4. Bon Opus Cat# CM71
Catalog No. 50-217-2822