Learn More
Bon Opus Biosciences Recombinant Mouse LEPR (C-mFc) -50ug
SDP
Supplier:  Bon Opus Biosciences C02M50UG
Encompass_Preferred
Recombinant Mouse Leptin Receptor is produced by our Mammalian expression system and the target gene encoding Leu22­Gly839 is expressed with a mFc tag at the C-terminus. Expression system: Human Cells. Accession# P48356. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# C02M
Catalog No. 50-208-0348