Learn More
Bon Opus Biosciences Recombinant Mouse LIF - Source: E. coli
SDP
Catalog No. NC2733779
Encompass_Preferred
Recombinant Mouse LIF is produced by our E. coli expression system and the target gene encoding Ser24-Phe203 is expressed. Accession#P09056. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.
Catalog No. NC2733779 Supplier Bon Opus Biosciences Supplier No. BP30750UG
Recombinant Mouse LIF is produced by our E. coli expression system and the target gene encoding Ser24-Phe203 is expressed. Accession#P09056. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.