Learn More
Bon Opus Biosciences Recombinant Mouse LILRB4 (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW5250UG
Encompass_Preferred
Recombinant Mouse Leukocyte Immunoglobulin-like Receptor Subfamily B Member 4 is produced by our Mammalian expression system and the target gene encoding Gly24-Lys238 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# Q64281. Formulation: Lyophilized from a 0.2 um filtered solution of 50 mM Tris-HCl,100mM Glycine,pH7.5. Bon Opus Cat# CW52
Catalog No. 50-208-0434