Learn More
Bon Opus Biosciences Recombinant Mouse Mesothelin (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CP6250UG
Encompass_Preferred
Recombinant Mouse Mesothelin is produced by our Mammalian expression system and the target gene encoding Asp298-Ser600 is expressed with a 6His tag at the C-terminus.  Expression system: Human Cells. Accession# Q61468. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# CP62
Catalog No. 50-208-0385