Learn More
Bon Opus Biosciences Recombinant Mouse Transcription factor A, mitochondrial/mTFAM (N-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP051500UG
Encompass_Preferred
Recombinant mouse mTFAM is produced by E.coli. The target gene encoding S43-H243 is expressed with a 6His tag at the N terminus. Expression system: E. coli. Accession# P40630. Formulation: Supplied as a 0.2 um filtered solution of 5umM TES, pH7.5. Bon Opus Cat# BP051
Catalog No. 50-208-0128