Learn More
Bon Opus Biosciences Recombinant Mouse NKG2D (N-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW9450UG
Encompass_Preferred
Recombinant Mouse NKG2-D Type II Integral Membrane Protein is produced by our Mammalian expression system and the target gene encoding Phe94 - Val232 is expressed with a Fc tag at the N-terminus. Expression system: Human Cells. Accession# O54709. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# CW94
Catalog No. 50-208-0471