Learn More
Bon Opus Biosciences Recombinant Mouse Notch 1 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CW3350UG
Encompass_Preferred
Recombinant Mouse Notch 1 is produced by our Mammalian expression system and the target gene encoding Ala18-Gln526 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q01705. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# CW33
Catalog No. 50-208-0419