Learn More
Bon Opus Biosciences Recombinant Mouse PDGF-A (N-FLAG) -500ug
SDP
Supplier:  Bon Opus Biosciences BP138500UG
Encompass_Preferred
Recombinant Mouse PDGF-A is produced by E.coli. The target gene encoding S87-T211 is expressed with a FLAG tag at the N-terminus. Expression system: E. coli. Accession# P20033. Formulation: Supplied as a 0.2 um filtered solution of PBS, um Urea pH 7.4. Bon Opus Cat# BP138
Catalog No. 50-208-0224