Learn More
Enzo Life Sciences Mu-Phe-hPhe-FMK (1mg)
SDP
Catalog No. 502010711
Encompass_Preferred
Irreversible, non-toxic, cell-permeable inhibitor of cysteine proteases calpain, cathepsins B and L, and Trypanosome trypanopain. Can be used in vitro (typically 20 µM) or in vivo. Inhibits tumor formation and growth. Alternative name: N-Morpholineurea-phenylalanyl-homophenylalanylfluoromethyl ketone. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25 mg/ml). Long Term Storage: +4°C.
Catalog No. 50-201-0711 Supplier Enzo Life Sciences Supplier No. BMLEI3230001
Irreversible, non-toxic, cell-permeable inhibitor of cysteine proteases calpain, cathepsins B and L, and Trypanosome trypanopain. Can be used in vitro (typically 20 µM) or in vivo. Inhibits tumor formation and growth. Alternative name: N-Morpholineurea-phenylalanyl-homophenylalanylfluoromethyl ketone. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25 mg/ml). Long Term Storage: +4°C.