Learn More
Bon Opus Biosciences Recombinant Mouse Siglec-15 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CW7150UG
Encompass_Preferred
Recombinant Mouse Sialic Acid-binding Ig-like Lectin 15 is produced by our Mammalian expression system and the target gene encoding Arg24-Thr262 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# A7E1W8. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,150mMnacl,5%Thehalose,0.3%chaps,Ph7.4. Bon Opus Cat# CW71
Catalog No. 50-208-0452