Learn More
Bon Opus Biosciences Recombinant Mouse TGFBR1 (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences C02H50UG
Encompass_Preferred
Recombinant Mouse TGF-beta Receptor Type-1 is produced by our Mammalian expression system and the target gene encoding Leu30-Glu125 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# Q64729. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# C02H
Catalog No. 50-208-0346