Learn More
Bon Opus Biosciences Recombinant Mouse Tissue Factor (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CU70500UG
Encompass_Preferred
Recombinant Mouse Tissue factor is produced by our Mammalian expression system and the target gene encoding Ala29-Glu251 is expressed with a 6His tag at the C-terminus. Accession# P20352. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM TrisHCl,150mM NaCl,0.05%Brij35,pH7.5. Bon Opus Cat# CU70
Catalog No. 50-217-2832