Learn More
Bon Opus Biosciences Recombinant Mouse TNF ligand superfamily member 9/TNFSF9(N-10His) 500ug
SDP
Supplier:  Bon Opus Biosciences CS34500UG
Encompass_Preferred
Recombinant Mouse 4-1BB Ligand is produced by our Mammalian expression system and the target gene encoding Arg104-Glu309 is expressed with a 10His tag at the N-terminus. Accession# P41274. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH7.4. Bon Opus Cat# CS34
Catalog No. 50-217-2829