Learn More
Bon Opus Biosciences Recombinant Mouse VSIG4 (C-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP02050UG
Encompass_Preferred
Recombinant Mouse V-Set and Ig Domain-Containing Protein 4 is produced by our Mammalian expression system and the target gene encoding His20-Pro187 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# F6TUL9. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP020
Catalog No. 50-208-0098