Learn More
Medchemexpress LLC HY-13501 100mg , Mubritinib TAK-165 CAS:366017-09-6 Purity:98%
SDP
Catalog No. 501493422
Encompass_Preferred
Medchemexpress, HY-13501 100mg Mubritinib TAK-165 CAS:366017-09-6 Formula:C25H23F3N4O2 IC50: 6 nM (EGFR2) Purity:98% Mubritinib (TAK-165) is a potent and selective EGFR2/HER2 inhibitor with an IC 50 of 6 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3422 Supplier Medchemexpress LLC Supplier No. HY13501100MG
Medchemexpress, HY-13501 100mg Mubritinib TAK-165 CAS:366017-09-6 Formula:C25H23F3N4O2 IC50: 6 nM (EGFR2) Purity:98% Mubritinib (TAK-165) is a potent and selective EGFR2/HER2 inhibitor with an IC 50 of 6 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.