Learn More
Selleck Chemical LLC Mubritinib (TAK 165) 200mg 366017-09-6
SDP
Catalog No. 506450
Encompass_Preferred
Mubritinib (TAK-165) is a potent inhibitor of HER2/ErbB2 with IC50 of 6 nM in BT-474 cell; no activity to EGFR, FGFR, PDGFR, JAK1, Src and Blk in BT-474 cell line. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-645-0 Supplier Selleck Chemical LLC Supplier No. S2216200MG
Mubritinib (TAK-165) is a potent inhibitor of HER2/ErbB2 with IC50 of 6 nM in BT-474 cell; no activity to EGFR, FGFR, PDGFR, JAK1, Src and Blk in BT-474 cell line. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.