Learn More
Medchemexpress LLC MUC-17 Human HEK29 50ug | 50ug
SDP
Catalog No. 5000194213
Encompass_Preferred
MUC-17 protein maintains mucosal homeostasis contributing to environmental stability Interacting via its C-terminus MUC-17 crucially associates with PDZK1 ensuring proper cellular localization MUC-17 Protein Human (HEK293 His) is the recombinant human-derived MUC-17 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-94213 Supplier Medchemexpress LLC Supplier No. HYP7874450UG
MUC-17 protein maintains mucosal homeostasis contributing to environmental stability Interacting via its C-terminus MUC-17 crucially associates with PDZK1 ensuring proper cellular localization MUC-17 Protein Human (HEK293 His) is the recombinant human-derived MUC-17 protein expressed by HEK293 with C-His labeled tag