Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4582MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: MUC1/845. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: MCF-7 or MDA-231 cells. Breast, colon, ovarian, endometrial carcinoma. Cellular Staining: Cytoplasmic, cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human milk-fat globule membranes (HMFGM). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 265-400kDa.Chromosome: 1q22. Gene Symbol: MUC1. Gene ID: 4582. Ensemble: ENSG00000185499. Uniprot: NP_001191226.
Catalog No. 50-188-4957