Learn More
Aviva Systems Biology Corp MUC1 Antibody - C-terminal region (ARP41445_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41445_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MUC1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human MUC1. Alternate names: CD227, EMA, PEM, PUM, mucin, KL-6, MAM6, PEMT, H23AG, MUC-1, MUC-1/X, MUC1/ZD, MUC-1/SECApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57858