Learn More
Aviva Systems Biology Corp MUC1 Antibody - middle region (ARP41456_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41456_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MUC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MUC1. Alternate names: CD227, EMA, H23AG, MAM6, PEM, PEMT, PUM, KL-6, MUC-1, MUC-1/X, MUC1/ZD, MUC-1/SECApplicationIHC, WB. Species reactivity: Human, Pig
Catalog No. 50-014-57865