Learn More
Enzo Life Sciences MUC1 Recombinant monoclonal antibody (rMUC1/960) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3680100
Encompass_Preferred
Clone: rMUC1/960. Isotype: IgG2bκ. Immunogen: Human milk fat globule membranes. Species reactivity: Human. Applications: Flow Cytometry, IF, IHC (PS). Purity: Protein G affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2896