Learn More
Enzo Life Sciences MUC1 Recombinant monoclonal antibody (HMFG2) (Rabbit IgG lambda) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6440200
Encompass_Preferred
Clone: HMFG2. Isotype: IgGλ. Immunogen: High-MW glycoprotein of the human milk fat globule. Species reactivity: Human. Applications: ELISA, IHC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3168