Learn More
Enzo Life Sciences MUC1 Recombinant monoclonal antibody (SM3) (Mouse IgG1 lambda) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6450200
Encompass_Preferred
Clone: SM3. Isotype: IgG1λ. Immunogen: Hydrogen fluoride deglycosylated milk mucin. Species reactivity: Human. Specificity: Recognizes the under-glycosylated form of human MUC1, a marker of tumors. Applications: ELISA, Flow Cytometry, IHC (FS), IHC (PS). Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3169