Learn More
Aviva Systems Biology Corp MUC15 Antibody - middle region (ARP45557_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45557_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MUC15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MUC15. Alternate names: PAS3, MUC-15, PASIIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-59051