Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4162MSM2XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM620. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: A-375, HUV-EC or HeLa Cells. Tonsil or Melanoma. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human MCAM protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 130kDa.Chromosome: 11q23.3. Gene Symbol: MCAM. Gene ID: 4162. Ensemble: ENSG00000076706. Uniprot: XP_016873251.
Catalog No. 50-188-4920