Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4583MSM1XXP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM513. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: LS174T cells or small intestine. Cellular Staining: Cytoplasmic, cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A synthetic peptide of 29 amino acids, KYPTTTPISTTTMVTPTPTPTGTQTPTTT from MUC2 protein, coupled to KLH. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 520kDa.Chromosome: 11p15.5. Gene Symbol: MUC2. Gene ID: 4583. Uniprot: NP_002448.
Catalog No. 50-188-4963