Learn More
Genetex Inc Anti-MUC2 antibody [C3], C-term
SDP
Supplier:  Genetex Inc GTX100664
Encompass
The Anti-MUC2 antibody [C3], C-term from GeneTex is a Rabbit Polyclonal antibody to MUC2. The Anti-MUC2 antibody [C3], C-term Recognizes Human, Mouse. The Anti-MUC2 antibody [C3], C-term has been validated for the following Applications: ICC/IF, IHC, IHC-P.
Catalog No. NC1598512