Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4584MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM200. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: LS174T cells or colon carcinoma, normal colon tissue. Cellular Staining: Cytoplasmic, cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A synthetic peptide of 35 amino acids, SIB35 (C-HSTPSFTSSITTTETTSHSTPSFTSSITTTETTS), which contains two of the MUC3 tandem repeats, coupled to KLH. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 1100kDa.Chromosome: 7q22.1. Gene Symbol: MUC3A. Gene ID: 57876. Ensemble: ENSG00000169894. Uniprot: NP_005951.
Catalog No. 50-188-4966