Learn More
Aviva Systems Biology Corp MUC3B Antibody - N-terminal region (ARP45578_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MUC3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MUC3B. Alternate names: MUC3, MUC3AApplicationWB. Species reactivity: Horse, Human, Rat
Catalog No. 50-014-59053