Learn More
Aviva Systems Biology Corp MUC3B Antibody - middle region (ARP58396_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58396_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MUC3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MUC3B. Alternate names: MUC3, MUC3AApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-014-62297