Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4586MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM297. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Stomach. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: >1,000kDa.Chromosome: 11p15.5. Gene Symbol: MUC5AC. Gene ID: 4586. Ensemble: ENSG00000283158. Uniprot: NP_001291288.
Catalog No. 50-188-4969