Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4588MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: CLH5. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Stomach. Cellular Staining: Cytoplasmic & Secreted. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A synthetic peptide of the Gastric Mucin 6 tandem repeat sequence. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 252kDa.Chromosome: 11p15.5. Gene Symbol: MUC6. Gene ID: 4588. Ensemble: ENSG00000277518. Uniprot: NP_005952.
Catalog No. 50-188-4975