Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ Mucasol™ Laboratory Detergent

Catalog No. 13688701 Shop All BrandTech Scientific Products
Change view
Click to view available options
Volume (Metric):
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-688-701 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-688-701 Supplier BRANDTECH™ Supplier No. 44003B

High performance, liquid alkaline cleaner that is non-toxic, non-aggressive, and biodegradable.

  • MUCASOL is a concentrated, alkaline cleaning agent for immersion and ultrasonic cleaning of laboratory glassware
  • Also suitable for porcelain, plastic, rubber, and metal
  • Can replace chromosulfuric acid
  • Free from chlorine and caustic alkalis
  • Not suitable for prolonged exposure to zinc or non-anodized aluminum surfaces
  • 2-liter bottle
  • Contains phosphates
TRUSTED_SUSTAINABILITY

Specifications

Product Type Alkaline Cleaning Agent
Color-coded No
Form Concentrate
pH 12.3
Product Line Mucasol
Scent Unscented
Sterility Non-sterile
Appearance Green
For Use With (Application) Cleaning laboratory glassware
Quantity 1 Ea.
Volume (Metric) 2 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.