Learn More
IMEB, Inc Muci-Carmine Stain Kit. Contains: 1-8 Oz. Bottle Metanil Yellow Stain, 1-8 Oz. Bottle Muci-Carmine (Stock), 1-8 Oz. Bottle Mayer’s Hematoxylin- Modified, 3-2 Oz. Staining Containers & Storage Tray.
SDP
Catalog No. 501689475
Encompass_Preferred
Designed to show Mucin and Capsule of crytococcus resulting in deep red with black nuclei. Contains: 1- 8 oz. Bottle Metanil Yellow Stain, 1- 8 oz. Bottle Muci-Carmine (Stock), 1- 8 oz. Bottle Mayer’s Hematoxylin- Modified, 3- 2 oz. Staining containers and storage tray.
Catalog No. 50-168-9475 Supplier IMEB, Inc Supplier No. K7398
Designed to show Mucin and Capsule of crytococcus resulting in deep red with black nuclei. Contains: 1- 8 oz. Bottle Metanil Yellow Stain, 1- 8 oz. Bottle Muci-Carmine (Stock), 1- 8 oz. Bottle Mayer’s Hematoxylin- Modified, 3- 2 oz. Staining containers and storage tray.