Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Mucic acid

Catalog No. M89617500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M89617500G 500 g
M89617100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M89617500G Supplier Sigma Aldrich Supplier No. M89617500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mucic acid

Specifications

CAS 526-99-8
Melting Point 220°C to 225°C (dec.) (lit.)
Molecular Formula C6H10O8
Linear Formula HOOC(CHOH)4 COOH
MDL Number MFCD00004239
Quantity 500 g
Synonym Galactaric acid; MTPA; Saccharolactic acid; Tetrahydroxyadipic acid; Tetrahydroxyhexanedioic acid
Molecular Weight (g/mol) 210.14
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.