Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mucic Acid, MP Biomedicals™

Catalog No. 0210237190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-102-37190 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-102-37190 Supplier MP Biomedicals, Inc Supplier No. 0210237190
Add to Cart
Add to Cart

Mucic Acid is also known as galactaric acid.

  • Hexaric acid from oxidative ring cleavage of galactose.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 526-99-8
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
InChI Key DSLZVSRJTYRBFB-UHFFFAOYNA-N
IUPAC Name 2,3,4,5-tetrahydroxyhexanedioic acid
SMILES OC(C(O)C(O)C(O)=O)C(O)C(O)=O

Specifications

Quantity 500 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.