Learn More
Diagnostic Biosystems MUCICARMINE Mucicarmine
Catalog No. 502407565
Encompass_Preferred
Mucicarmine Stain Kit is intended for use in the histological visualization of acid mucopolysacharides in tissue sections. This product is useful in distinguishing mucin negative undifferentiated squamous cell lesions from mucin positive adenocarcinomas. In addition, this product will stain the mucopolysaccharide capsule of Cryptococcus neoformans.
Catalog No. 50-240-7565 Supplier Diagnostic Biosystems Supplier No. KT024
Mucicarmine Stain Kit is intended for use in the histological visualization of acid mucopolysacharides in tissue sections. This product is useful in distinguishing mucin negative undifferentiated squamous cell lesions from mucin positive adenocarcinomas. In addition, this product will stain the mucopolysaccharide capsule of Cryptococcus neoformans.