Learn More
Newcomer Supply Mucicarmine Stock Stain, Mayer, 125 ml
SDP
Catalog No. NC9066494
Encompass
Newcomer Supply Mucicarmine Stock Stain, Mayer a key element in the Mucin, Mayer Mucicarmine Stain procedure, is used to stain acid epithelial mucin (sialomucin, sulfomucin) and also useful for the demonstration of the encapsulated yeast Cryptococcus neoformans. The Metanil Yellow or Tartrazine Stain counterstain will lend a light yellow color to connective tissue and cytoplasmic elements. Shelf Life is 1 year from date of manufacture. Store at 2-8°C.
Catalog No. NC9066494 Supplier Newcomer Supply Supplier No. 1250A
Newcomer Supply Mucicarmine Stock Stain, Mayer a key element in the Mucin, Mayer Mucicarmine Stain procedure, is used to stain acid epithelial mucin (sialomucin, sulfomucin) and also useful for the demonstration of the encapsulated yeast Cryptococcus neoformans. The Metanil Yellow or Tartrazine Stain counterstain will lend a light yellow color to connective tissue and cytoplasmic elements. Shelf Life is 1 year from date of manufacture. Store at 2-8°C.