Learn More
Mucicarmine Stock Solution, 250 mL, G-Biosciences
SCI_ED
Catalog No. S001137
Catalog No. S001137 Supplier G-Biosciences Supplier No. 7862240
Add to Cart
Add to Cart
Mucicarmine stock solution is used for the histological staining of tissue mucins with epithelial origin.
  • It is used to distinguish mucin negative undifferentiated squamous cell lesions from mucin positive adenocarcinomas. In addition, it can stain the mucopolysaccharide capsule of Cryptococcus neoformans.
Specifications
Product Type Mucicarmine Stock Solution