Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mucicarmine, TCI America™
SDP

Catalog No. M046825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M046825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M046825G Supplier TCI America Supplier No. M046825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51395-97-2
MDL Number MFCD00071289

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Mucicarmine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.