Learn More
ACROBiosystems ACROBiosystems Human Mucin-1 / MUC-1 (890-1158) Protein, His Tag
Catalog No. 501956613
Encompass_Preferred
Recombinant Protein;100UG;Human Mucin-1 (890-1158), His Tag (MU1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Ser 890 - Gly 1158 (Accession # P15941-1).;This protein carries a polyhistidine tag at the C-terminus. The mature form of Mucin-1 is a non-covalent heterodimeric complex with the proteolytically cleaved partial α and partial β chain. Each partial α and partial β chain has a calculated MW of 21.5 kDa (partial α chain) and 8.4 kDa (partial β chain). The protein migrates as 45-80 kDa (partial α chain) and 10 kDa (partial β chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6613 Supplier ACROBiosystems Supplier No. MU1H52H9100UG
Recombinant Protein;100UG;Human Mucin-1 (890-1158), His Tag (MU1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Ser 890 - Gly 1158 (Accession # P15941-1).;This protein carries a polyhistidine tag at the C-terminus. The mature form of Mucin-1 is a non-covalent heterodimeric complex with the proteolytically cleaved partial α and partial β chain. Each partial α and partial β chain has a calculated MW of 21.5 kDa (partial α chain) and 8.4 kDa (partial β chain). The protein migrates as 45-80 kDa (partial α chain) and 10 kDa (partial β chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.