Learn More
ACROBiosystems ACROBiosystems Human Mucin-1 / MUC-1 (1036-1155) Protein, His Tag
Catalog No. 501956603
Encompass_Preferred
Recombinant Protein;100UG;Human Mucin-1 (1036-1155), His Tag (MU1-H52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 1036 - Gly 1155 (Accession # P15941-1).;This protein carries a polyhistidine tag at the C-terminus. The mature form of Mucin-1 is a non-covalent heterodimeric complex with the proteolytically cleaved partial α and partial β chain. Each partial α and partial β chain has a calculated MW of 7.2 kDa (partial α chain) and 8.2 kDa (partial β chain). The protein migrates as 13-15 kDa (partial α chain) and 10 kDa (partial β chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6603 Supplier ACROBiosystems Supplier No. MU1H52H5100UG
Recombinant Protein;100UG;Human Mucin-1 (1036-1155), His Tag (MU1-H52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 1036 - Gly 1155 (Accession # P15941-1).;This protein carries a polyhistidine tag at the C-terminus. The mature form of Mucin-1 is a non-covalent heterodimeric complex with the proteolytically cleaved partial α and partial β chain. Each partial α and partial β chain has a calculated MW of 7.2 kDa (partial α chain) and 8.2 kDa (partial β chain). The protein migrates as 13-15 kDa (partial α chain) and 10 kDa (partial β chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.