Learn More
ACROBiosystems ACROBiosystems Human Mucin-1 / MUC-1 Protein, Llama IgG2b Fc Tag, low endotoxin
Catalog No. 501956776
Encompass_Preferred
Recombinant Protein;1MG;Human Mucin-1, Llama IgG2b Fc Tag, low endotoxin (MU1-H5255) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Gly 167 (Accession # AAI20976).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 42.4 kDa. The protein migrates as 50-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6776 Supplier ACROBiosystems Supplier No. MU1H52551MG
Recombinant Protein;1MG;Human Mucin-1, Llama IgG2b Fc Tag, low endotoxin (MU1-H5255) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Gly 167 (Accession # AAI20976).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 42.4 kDa. The protein migrates as 50-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.