Learn More
Biomatik Corporation Mucin 2 (Muc2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU31196
Encompass_Preferred
Mucin 2 (Muc2) Polyclonal Antibody is a Pig (Sus scrofa; Porcine) Polyclonal Antibody to Mucin 2 (MUC2). The Mucin 2 (Muc2) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Pig (Sus scrofa; Porcine). Immunogen: Val36~Cys351. Applications: WB, IHC. Target Synonyms: Mucin 2 (MUC2). Accession Number: XP_013845199.1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6730