Learn More
Biomatik Corporation Mucin 5 Subtype B (Muc5B) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29286
Encompass_Preferred
Mucin 5 Subtype B (Muc5B) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Mucin 5 Subtype B (MUC5B). The Mucin 5 Subtype B (Muc5B) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Cys5653~Val5762. Applications: WB. Target Synonyms: Mucin 5 Subtype B (MUC5B). Accession Number: Q9HC84. Gene ID: 727897. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7573