Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Mucin, Bovine Submaxillary Gland

Catalog No. AAJ63859MC
Change view
Click to view available options
Quantity:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63859MC 100mg
AAJ63859ME 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63859MC Supplier Thermo Scientific Chemicals Supplier No. J63859MC
Add to Cart

missing translation for 'validMainframeMsg'

Mucin, CAS # 84195-52-8, is a high molecular weight glycoprotein, and is a major constituent of saliva, gastrointestinal juices, and other secretions. It is used as a substrate for the viral enzyme neuraminidase. | 400-4000KDa

Mucin is used in a study to investigate a galactose-specific lectin from the red marine alga Ptilota filicina. Mucin from bovine submaxillary glands has also been used in a study to investigate the characterization of a unique mucin-like glycoprotein secreted by a human endometrial adenocarcinoma cell line, neuraminidase substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Mucin derived from the bovine submaxillary gland is a glycoprotein present in saliva, gastrointestinal juices, and other secretions that promote cilia lubrication

Application

  • Mucin can be used as a source of O-linked oligosaccharides for the viral enzyme, neuraminidase
  • Mucins are glycoproteins with roles in immunity and digestion
  • In biological research, mucin is used to investigate a galactose-specific lectin from the red marine alga Ptilota filicina
  • It is used to investigate the characterization of a unique mucin-like glycoprotein secreted by a human endometrial adenocarcinoma cell line
  • It is used as a substrate for recombinant sialidase Gardnerella vaginalis
  • It is used in protocols to quantify mucin
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Neuraminidase Substrate
Formulation Lyophilized powder
Molecular Weight (g/mol) 400-4000KDa
Quantity 100mg
Source Bovine submaxillary gland
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Mucin, Bovine submaxillary gland
Recombinant Native
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.